missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, Optima™LC/MS Grade, Fisher Chemical™

Catalog Number 10091304
41.56 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.


Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
67-63-0
C3H8O
60.096
11674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
11674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O

2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm

TRUSTED_SUSTAINABILITY
pH 7
Melting Point -89.5°C
Absorbance Report AU at 210nm, Report AU at 220nm, Report AU at 230nm, Report AU at 254nm
Color Colorless
Density 0.785g/cm³
Boiling Point 81°C
Quantity 1 L
Identification Pass Test
Packaging Wide Neck Amber Bottle
Physical Form Liquid
Assay Percent Range Assay: ≥99.90%
Residue after Evaporation 1ppm max.
Linear Formula (CH3)2CHOH
Vapor Pressure 43mmHg at 20°C
Viscosity 2.27 mPaS at 20°C
Water 0.05% max.
Merck Index 15, 5254
Formula Weight 60.1
Percent Purity ≥99.90%
Grade Also meets ACS specifications
Chemical Name or Material 2-Propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iso-propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'