Learn More
Invitrogen™ MgCl2 (1 M)
Molecular biology grade, Invitrogen 1M MgCl2 solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use.
Brand: Invitrogen™ AM9530G
41.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : 50
Additional Details : Weight : 0.10500kg
Description
Molecular biology grade, Invitrogen 1M MgCl2 solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature
Specifications
Magnesium Chloride | |
1 M | |
Ambion™ | |
100 mL | |
Liquid |
Store at room temperature. | |
RNA Applications | |
Molecular Biology Grade | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.