Learn More
Corning™ Transwell™ Multiple Well Plate with Permeable Polyester Membrane Inserts
GREENER_CHOICEBIOPHARMA
Featuring a microscopically transparent polyester membrane that is TC-treated for optimal cell attachment and growth. Corning™ Transwell™ Multiwell Plate with Polyester Membrane Inserts provide better visibility under phase contrast microscopy and allow assessment of cell viability and monolayer formation.
£215.00 - £399.00
Specifications
Surface Coating TC-Treated
Color Clear
Surface Treatment TC-Treated
Sterility Sterile
View More Specs
Products 7
Product Code Brand Pore Size Description Culture Area Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Pore Size Description Culture Area Diameter (Metric) Outer Price Quantity & Availability  
10317701
GREENER_CHOICEBIOPHARMA
View Documents
Corning™
3472
3 μm 12 inserts in a 24-well plate 0.33 cm2 6.5 mm
£370.00
Pack of 48
In Stock
to see stock.
Add to basket
10482181
GREENER_CHOICEBIOPHARMA
View Documents
Corning™
3470
0.4 μm 12 inserts in a 24-well plate 0.33 cm2 6.5 mm
£370.00
Pack of 48
In Stock
to see stock.
Edge
Add to basket
13216768
GREENER_CHOICEBIOPHARMA
View Documents
Corning™
3464
8 μm 12 inserts in a 24-well plate 0.33 cm2 6.5 mm
£399.00
Pack of 48
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
10357651
GREENER_CHOICEBIOPHARMA
View Documents
Corning™
3462
3 μm 12 inserts in a 12-well plate 1.12 cm2 12 mm
£370.00
Pack of 48
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
10565482
GREENER_CHOICEBIOPHARMA
View Documents
Corning™
3460
0.4 μm 12 inserts in a 12-well plate 1.12 cm2 12 mm
£370.00
Pack of 48
In Stock
to see stock.
Edge
Add to basket
10619141
GREENER_CHOICEBIOPHARMA
View Documents
Corning™
3450
0.4 μm 6 inserts in a 6-well plate 4.67 cm2 24 mm
£215.00
Pack of 24
In Stock
to see stock.
Edge
Add to basket
10699141
GREENER_CHOICEBIOPHARMA
View Documents
Corning™
3452
3 μm 6 inserts in a 6-well plate 4.67 cm2 24 mm
£215.00
Pack of 24
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
Description
Description
  • Ideal for transport studies, chemotaxis, co-culture, and microbial pathogenesis studies
  • Thin membrane and straight pore structure provides consistent transport and cell migration
  • Self-centering insert prevents media from wicking between the insert and the outer well
  • Plate: Virgin Polystyrene
  • Membrane: PET
Specifications
Specifications
TC-Treated
Clear
TC-Treated
Sterile