Learn More
Thermo Scientific™ FastDigest BamHI
Cut at G^GATCC sites with Thermo Scientific™ FastDigest™ BamHI restriction enzyme, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer.
£42.45 - £118.00
Specifications
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme BamH I
Compatible Buffer 10X FastDigest Buffer/FastDigest Green Buffer
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10334480
View Documents Promotion Details
Thermo Scientific™
FD0054
800 μL (800 Reactions)
£42.45
800 reactions
11873833
View Documents Promotion Details
Thermo Scientific™
FD0055
5 x 500 μL (2,500 Reactions)
£118.00
2500 reactions
Description
Show More
Specifications
Specifications
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
10X FastDigest Buffer/FastDigest Green Buffer
37°C
Restriction Enzyme
FastDigest™
80°C
BamH I
Yes
No
Traditional Cloning
5' protruding ends
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Thermo Scientific™ FastDigest BamHI