Learn More
Fisherbrand™ Wash Bottles with Adjustable Sprayer
Handle hot/cold, aqueous/solvent-based solutions
£37.90 - £76.00
Specifications
Includes Polypropylene adjustable sprayer
Material HDPE
Sterility Non-sterile
Shape Squat
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11714356
View Documents
Fisherbrand™
90010-81
240 mL
£37.90
Pack of 3
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
11795873
View Documents
Fisherbrand™
90023-74
1000 mL
£76.00
Each
Estimated Shipment: 11-03-2025
to see stock.
Add to basket
Description
Description
Handle hot/cold, aqueous/solvent-based solutions using Fisherbrand™ Wash Bottles with Adjustable Sprayer.
Fully adjustable spray pattern: “fine mist” for TLC plates; “needle” for rinsing glassware; “big stream” for powerful jet needed to wash filters. 
  • Bottle is made of high-density polyethylene
  • Sprayer is polypropylene
  • Trigger allows one-handed, controlled delivery with light pressure
  • Engraved markings show how much fluid is left
  • Squat shape takes up little bench space and ensures a low center of gravity
Specifications
Specifications
Polypropylene adjustable sprayer
HDPE
Non-sterile
Squat