Learn More
Kimtech Science™ Wipe Delicate Task White
Wipe liquids with multi-ply absorbency and low lint content for intricate dusting. Kimberly Clark™ Kimtech Scientific™ Delicate Task Wipers are ideal for cleaning delicate glassware, instruments, and surface/parts.
Brand:  Kimtech 7557
53.78 GBP valid until 2024-05-31
Use promo code "21679" to get your promotional price.
Additional Details : Weight : 3.41200kg
Product Code. 17701868
  • £119.00 / Pack of 2400
In Stock
to see stock.
Add to basket
Description
Description
A range of high-performance wipers, suitable for ISO Class 4 or higher laboratory environments.
Ideal for: cleaning of delicate glassware, instruments and surface/parts; wiping liquids, with multi-ply absorbency and low lint content for intricate dusting.
Available as: 100 or 200 format, white paper wipers, dispensed from a POP-UP Box flexible system, giving controlled access to hygienically protected wipers when you’re cleaning or wiping on the move.
Specifications
Specifications
Wipe
21 cm
White
Suitable for ISO Class 4 or higher laboratory environments.
2-ply
20 cm
Paper
Wiping liquids or cleaning delicate glass, instruments, parts, and surfaces.
100 Sheets x 24 Dispenser Boxes