Learn More
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
£23.10 - £420.00
Chemical Identifiers
CAS | 128-08-5 |
---|---|
MDL Number | MFCD00005510 |
InChI Key | PCLIMKBDDGJMGD-UHFFFAOYSA-N |
Synonym | n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide |
PubChem CID | 67184 |
ChEBI | CHEBI:53174 |
IUPAC Name | 1-bromopyrrolidine-2,5-dione |
SMILES | C1CC(=O)N(C1=O)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10559820
|
Thermo Scientific Acros
107450100 |
10 g | Glass bottle |
£27.90
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10655332
|
Thermo Scientific Acros
107451000 |
100 g | Glass bottle |
£23.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10478820
|
Thermo Scientific Acros
107455000 |
500 g | Glass bottle |
£79.20
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10132900
|
Thermo Scientific Acros
107450010 |
1 kg | Glass bottle |
£121.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11925581
|
Thermo Scientific Acros
107450050 |
5 kg | Plastic drum |
£420.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent, C-X bond formation reagentChemical Identifiers
128-08-5 | |
PCLIMKBDDGJMGD-UHFFFAOYSA-N | |
67184 | |
1-bromopyrrolidine-2,5-dione |
MFCD00005510 | |
n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide | |
CHEBI:53174 | |
C1CC(=O)N(C1=O)Br |
Specifications
128-08-5 | |
100.0 | |
White to Yellow | |
1% max. | |
98.5 to 101.5% (on dry substance) (Iodometry) | |
MFCD00005510 | |
01,78; 02,40; 03,34; 04,49; 05,65; 06,74; 07,37; 08,54; 09,70; 10,57; 11,79; 12,79; 13,41; 14,57; 15,50; 16,49 | |
n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide | |
PCLIMKBDDGJMGD-UHFFFAOYSA-N | |
1-bromopyrrolidine-2,5-dione | |
67184 | |
177.99 | |
Crystalline Powder |
98.5 | |
175.0°C to 178.0°C | |
180°C | |
Authentic | |
C4H4BrNO2 | |
21, 380 | |
15, 1448 | |
Solubility in water: 14.8g/L (20°C). Other solubilities: 3.10g/100g glacial acetic acid(25°C),g/100g solvent (25°C):,tert. butanol:0.73- aceton:14.40,carbone tetrachloride:0.02- hexane:0.006,benzene:0.5 | |
C1CC(=O)N(C1=O)Br | |
177.99 | |
CHEBI:53174 | |
99% | |
N-Bromosuccinimide |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
In case of major fire and large quantities: Evacuate area.
Fight fire rem
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-877-2
missing translation for 'tsca' : TSCA
- N-Bromosuccinimide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only