Learn More
Contec™ NeutraKlean, 1L
Contec™ NeutraKlean is a gentle and low-foaming detergent designed for the cleaning of life science cleanrooms.
Brand: Contec™ FBT103NK
72.35 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : CAS Number : 2605-79-0 Weight : 6.00000kg
Description
- Neutral pH makes it suitable for use in all areas of cleanroom as gentle on surfaces.
- Filtered to 0.2 microns under Grade A airflow in an Grade C cleanroom ensures the product is free from contamination and particulates
- Low-foaming detergent action is ideal for use on floors, walls, and benches
- Low level of surfactant provides low residue which can be easily removed
- Trigger spray can be set to jet or spray and provides good surface coverage
- Trigger spray and bag in bottle protected system prevents bottle contents from becoming contaminated during use
- Bottles can be completely emptied eliminating wastage so no need to discard unused product during shelf life
Specifications
Non-sterile | |
No | |
6/Cs. | |
1 L |