man

Test

CAS RN 1003-68-5

CH3NHO

CAS RN 1003-68-5

IUPAC Name: 5-methyl-1,2-dihydropyridin-2-one
Synonyms: 5-methyl-1H-pyridin-2-one
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: SOHMZGMHXUQHGE-UHFFFAOYSA-N
SMILES: CC1=CNC(=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results