man

Test

CAS RN 1008997-34-9

H3COOHNHSOOCl

CAS RN 1008997-34-9

IUPAC Name: 2-(3-chlorobenzenesulfonamido)propanoic acid
Synonyms: 2-(3-chlorobenzenesulfonamido)propanoic acid
Molecular Weight (g/mol): 263.69
Molecular Formula: C9H10ClNO4S
InChi Key: LKWSJYWATSRWQH-UHFFFAOYNA-N
SMILES: CC(NS(=O)(=O)C1=CC(Cl)=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)