CAS RN 1015082-78-6
CAS RN 1015082-78-6
IUPAC Name:
potassium trifluoro[4-(hydroxymethyl)phenyl]boranuide
Synonyms:
potassium trifluoro[4-(hydroxymethyl)phenyl]boranuide
Molecular Weight (g/mol):
214.04
Molecular Formula:
C7H7BF3KO
InChi Key:
HQOSFYRRFPFWTG-UHFFFAOYSA-N
SMILES:
[K+].OCC1=CC=C(C=C1)[B-](F)(F)F
1
–
1
of
1
results
1
Potassium 4-(hydroxymethyl)phenyltrifluoroborate, 95%, Thermo Scientific™
CAS: 1015082-78-6 Molecular Formula: C7H7BF3KO Molecular Weight (g/mol): 214.036 MDL Number: MFCD08276800 InChI Key: HQOSFYRRFPFWTG-UHFFFAOYSA-N Synonym: potassium 4-hydroxymethyl phenyltrifluoroborate,potassium 4-hydroxymethylphenyl ltrifluoroborate,potassium trifluoro 4-hydroxymethyl phenyl boranuide,pubchem11367,amtb228,potassium 4-hydroxymethyl phenyl ltrifluoroborate,potassium trifluoro 4-hydroxymethyl phenyl borate,potassium trifluoro 4-hydroxymethyl phenyl borate 1- PubChem CID: 23697349 IUPAC Name: potassium;trifluoro-[4-(hydroxymethyl)phenyl]boranuide SMILES: [B-](C1=CC=C(C=C1)CO)(F)(F)F.[K+]