man

Test

CAS RN 1017779-71-3

H3COH2NFF

CAS RN 1017779-71-3

IUPAC Name: 4,5-difluoro-2-methoxyaniline
Synonyms: 4,5-difluoro-2-methoxyaniline
Molecular Weight (g/mol): 159.14
Molecular Formula: C7H7F2NO
InChi Key: JGCKFBTYRYSVJG-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(F)C=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results