man

Test

CAS RN 10250-48-3

H3COOCH3CH3H3C

CAS RN 10250-48-3

IUPAC Name: methyl 3,3-dimethylbutanoate
Synonyms: methyl 3,3-dimethylbutanoate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: DXBOTVWRXLQVMG-UHFFFAOYSA-N
SMILES: COC(=O)CC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results