man

Test

CAS RN 103-03-7

H2NOHNNH

CAS RN 103-03-7

IUPAC Name: (phenylamino)urea
Synonyms: phenylaminourea
Molecular Weight (g/mol): 151.17
Molecular Formula: C7H9N3O
InChi Key: AVKHCKXGKPAGEI-UHFFFAOYSA-N
SMILES: NC(=O)NNC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results