man

Test

CAS RN 103-94-6

HOOONHNOO

CAS RN 103-94-6

IUPAC Name: [(4-nitrophenyl)carbamoyl]formic acid
Synonyms: 4-nitrophenyloxamic acid
Molecular Weight (g/mol): 210.15
Molecular Formula: C8H6N2O5
InChi Key: ZEJKSYPGUAUQKW-UHFFFAOYSA-N
SMILES: OC(=O)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results