man

Test

CAS RN 10341-26-1

OHNO

CAS RN 10341-26-1

IUPAC Name: 1,4-oxazepan-5-one
Synonyms: 1,4-oxazepan-5-one
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: OXDIYMHNQAWSCL-UHFFFAOYSA-N
SMILES: O=C1CCOCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results