man

Test

CAS RN 10361-84-9

ClClClSc

CAS RN 10361-84-9

IUPAC Name: scandium(3+) trichloride
Synonyms: scandium chloride
Molecular Weight (g/mol): 151.31
Molecular Formula: Cl3Sc
InChi Key: DVMZCYSFPFUKKE-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Sc+3]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results