man

Test

CAS RN 104197-13-9

OHFBrF

CAS RN 104197-13-9

IUPAC Name: 4-bromo-2,6-difluorophenol
Synonyms: 4-bromo-2,6-difluorophenol
Molecular Weight (g/mol): 208.99
Molecular Formula: C6H3BrF2O
InChi Key: GPRPSJPFAAGLCA-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(Br)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results