man

Test

CAS RN 104594-70-9

HOOOHO

CAS RN 104594-70-9

IUPAC Name: 2-phenylethyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
Synonyms: caffeic acid phenethyl ester capeee cape compound
Molecular Weight (g/mol): 284.31
Molecular Formula: C17H16O4
InChi Key: SWUARLUWKZWEBQ-VQHVLOKHSA-N
SMILES: OC1=CC=C(\C=C\C(=O)OCCC2=CC=CC=C2)C=C1O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results