man

Test

CAS RN 1048970-16-6

CH3HNO

CAS RN 1048970-16-6

IUPAC Name: [(3,4-dihydro-2H-1-benzopyran-8-yl)methyl](methyl)amine
Synonyms: (3,4-dihydro-2H-1-benzopyran-8-ylmethyl)(methyl)amine
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: VVCGFZWBAXPSIO-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC2=C1OCCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results