man

Test

CAS RN 105-43-1

H3CCH3OHO

CAS RN 105-43-1

IUPAC Name: 3-methylpentanoic acid
Synonyms: 3-methyl valeric acid
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: IGIDLTISMCAULB-UHFFFAOYNA-N
SMILES: CCC(C)CC(O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results