man

Test

CAS RN 10522-47-1

H3COBrN

CAS RN 10522-47-1

IUPAC Name: 5-bromo-8-methoxyquinoline
Synonyms: 5-bromo-8-methoxyquinoline
Molecular Weight (g/mol): 238.08
Molecular Formula: C10H8BrNO
InChi Key: ZRQISUREPLHYIG-UHFFFAOYSA-N
SMILES: COC1=C2N=CC=CC2=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results