man

Test

CAS RN 10531-43-8

SOBr

CAS RN 10531-43-8

IUPAC Name: 2-bromo-1-(5-phenylthiophen-2-yl)ethan-1-one
Synonyms: 2-bromo-1-(5-phenylthiophen-2-yl)ethanone
Molecular Weight (g/mol): 281.17
Molecular Formula: C12H9BrOS
InChi Key: BOIAFAJKOBVLGS-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C(S1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results