man

Test

CAS RN 10538-49-5

CH3OOH3COHO

CAS RN 10538-49-5

IUPAC Name: 3-(2,5-dimethoxyphenyl)propanoic acid
Synonyms: 3-(2,5-dimethoxyphenyl)propanoic acid
Molecular Weight (g/mol): 210.23
Molecular Formula: C11H14O4
InChi Key: JENQUCZZZGYHRW-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(CCC(O)=O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results