man

Test

CAS RN 10541-56-7

H3CCH3O

CAS RN 10541-56-7

IUPAC Name: 1-(4-octylphenyl)ethan-1-one
Synonyms: 1-(4-octylphenyl)ethanone
Molecular Weight (g/mol): 232.37
Molecular Formula: C16H24O
InChi Key: GARQDIVXKVBJFP-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(C=C1)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results