man

Test

CAS RN 10553-11-4

NOOHNO

CAS RN 10553-11-4

IUPAC Name: 4-nitro-1H-indole-3-carbaldehyde
Synonyms: 4-nitro-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 190.16
Molecular Formula: C9H6N2O3
InChi Key: CGXVTWQTGQAMMX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2C(NC=C2C=O)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results