man

Test

CAS RN 105946-82-5

IBr

CAS RN 105946-82-5

IUPAC Name: 4-bromo-4'-iodo-1,1'-biphenyl
Synonyms: 4-bromo-4'-iodo-1,1'-biphenyl
Molecular Weight (g/mol): 359
Molecular Formula: C12H8BrI
InChi Key: GWOAJJWBCSUGHH-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results