man

Test

CAS RN 106-25-2

CH3H3CCH3OH

CAS RN 106-25-2

IUPAC Name: (2Z)-3,7-dimethylocta-2,6-dien-1-ol
Synonyms: nerol (E)3,7-dimethyl-octadien-1-ol vernol nerol (natural) guaniol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: GLZPCOQZEFWAFX-YFHOEESVSA-N
SMILES: CC(C)=CCC\C(C)=C/CO
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)