man

Test

CAS RN 106-35-4

H3COCH3

CAS RN 106-35-4

IUPAC Name: heptan-3-one
Synonyms: 3-heptanone etilbutilchetone eptan-3-one ethyl-n-butyl ketone ethylbutylcetone aethylbutylketon
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: NGAZZOYFWWSOGK-UHFFFAOYSA-N
SMILES: CCCCC(=O)CC
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)