man

Test

CAS RN 106-40-1

NH2Br

CAS RN 106-40-1

IUPAC Name: 4-bromoaniline
Synonyms: 4-bromoaniline P-bromoaniline
Molecular Weight (g/mol): 172.03
Molecular Formula: C6H6BrN
InChi Key: WDFQBORIUYODSI-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C=C1
Boiling Point 
  • (3)
  • (6)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)