CAS RN 106-94-5
CAS RN 106-94-5
IUPAC Name:
1-bromopropane
Synonyms:
1-bromopropane
Molecular Weight (g/mol):
122.99
Molecular Formula:
C3H7Br
InChi Key:
CYNYIHKIEHGYOZ-UHFFFAOYSA-N
SMILES:
CCCBr
1
–
1
of
1
results
1
1-Bromopropane, 99%, Thermo Scientific Chemicals
CAS: 106-94-5 Molecular Formula: C3H7Br MDL Number: MFCD00000254 InChI Key: CYNYIHKIEHGYOZ-UHFFFAOYSA-N Synonym: n-propyl bromide,propyl bromide,propane, 1-bromo,propane, bromo,bromopropane,1-bromo-propane,1-propyl bromide,ccris 30,unii-y9746dne68,n-propylbromide PubChem CID: 7840 ChEBI: CHEBI:47105 IUPAC Name: 1-bromopropane SMILES: CCCBr