man

Test

CAS RN 1060805-03-9

H3CONF

CAS RN 1060805-03-9

IUPAC Name: 3-fluoro-4-methoxypyridine
Synonyms: 3-fluoro-4-methoxypyridine
Molecular Weight (g/mol): 127.12
Molecular Formula: C6H6FNO
InChi Key: XYTUUTJNWFECNS-UHFFFAOYSA-N
SMILES: COC1=C(F)C=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results