man

Test

CAS RN 106224-36-6

CH3CH3H3COOCH3H3CCH3OOPd

CAS RN 106224-36-6

IUPAC Name: palladium(2+) bis(2,2-dimethylpropanoate)
Synonyms: palladium(2+) dipivalate
Molecular Weight (g/mol): 308.67
Molecular Formula: C10H18O4Pd
InChi Key: PAGZTSLSNQZYEV-UHFFFAOYSA-L
SMILES: [Pd++].CC(C)(C)C([O-])=O.CC(C)(C)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results