CAS RN 1066-45-1
CAS RN 1066-45-1
IUPAC Name:
trimethyltin chloride
Synonyms:
trimethyltin chloride
Molecular Weight (g/mol):
201.28
Molecular Formula:
C3H11ClSn
InChi Key:
FVFGWISLDLUGHX-UHFFFAOYSA-N
SMILES:
Cl.C[SnH](C)C
1
–
3
of
3
results
2
Trimethyltin chloride, Thermo Scientific Chemicals
CAS: 1066-45-1 Molecular Formula: C3H11ClSn Molecular Weight (g/mol): 201.28 MDL Number: MFCD00000520 InChI Key: FVFGWISLDLUGHX-UHFFFAOYSA-N Synonym: trimethyltin chloride,stannane, chlorotrimethyl,chlorotrimethyltin,trimethylchlorotin,trimethylstannyl chloride,trimethylchlorostannane,chloro trimethyl stannane,m&t chemicals 1222-45,stannylium, trimethyl-, chloride,trimethyltinchloride PubChem CID: 14016 IUPAC Name: chloro(trimethyl)stannane SMILES: Cl.C[SnH](C)C