man

Test

CAS RN 106614-28-2

CH3OOFF

CAS RN 106614-28-2

IUPAC Name: methyl 2,4-difluorobenzoate
Synonyms: methyl 2,4-difluorobenzoate
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: YQUHULOBTDYMAG-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(F)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results