man

Test

CAS RN 106877-33-2

NH2NFFF

CAS RN 106877-33-2

IUPAC Name: 6-(trifluoromethyl)pyridin-3-amine
Synonyms: 6-(trifluoromethyl)pyridin-3-amine
Molecular Weight (g/mol): 162.12
Molecular Formula: C6H5F3N2
InChi Key: IBOZOWZSXZNIHI-UHFFFAOYSA-N
SMILES: NC1=CC=C(N=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results