man

Test

CAS RN 107-43-7

H3CNH3CCH3OO

CAS RN 107-43-7

IUPAC Name: 2-(trimethylazaniumyl)acetate
Synonyms: Bet abromine betafin bcr citrate de bétaïne upsa scorbo bétaïne betafin betaine glycylbetaine loramine amb 13 aminocoat
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: KWIUHFFTVRNATP-UHFFFAOYSA-N
SMILES: C[N+](C)(C)CC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)