man

Test

CAS RN 107-49-3

CH3OPOOCH3OPOOCH3OCH3

CAS RN 107-49-3

IUPAC Name: diethyl [(diethoxyphosphoryl)oxy]phosphonate
Synonyms: tetraethyl pyrophosphate
Molecular Weight (g/mol): 290.19
Molecular Formula: C8H20O7P2
InChi Key: IDCBOTIENDVCBQ-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)OP(=O)(OCC)OCC
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
For Use With (Application) 
  • (1)
For Use With (Equipment) 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)