CAS RN 1071-83-6
CAS RN 1071-83-6
IUPAC Name:
2-[(phosphonomethyl)amino]acetic acid
Synonyms:
glyphosate epa pesticide chemical code 417300 glyphosate, dilithium salt magnesium salt (2:1) glyphosate glyphosate, zinc salt roundup max calcium salt glyphosate magnesium salt glyphosate monosodium salt glyphosate glyphosate, monopotassium salt
Molecular Weight (g/mol):
169.07
Molecular Formula:
C3H8NO5P
InChi Key:
XDDAORKBJWWYJS-UHFFFAOYSA-N
SMILES:
OC(=O)CNCP(O)(O)=O
1
–
7
of
7
results
1
Thermo Scientific Chemicals Glyphosate
CAS: 1071-83-6 Molecular Formula: C3H8NO5P Molecular Weight (g/mol): 169.07 MDL Number: MFCD00055350 InChI Key: XDDAORKBJWWYJS-UHFFFAOYSA-N IUPAC Name: 2-[(phosphonomethyl)amino]acetic acid SMILES: OC(=O)CNCP(O)(O)=O
6
Glyphosate, Fluka™
Analytical standard, reference material