man

Test

CAS RN 1072-15-7

NaH3COSOOO

CAS RN 1072-15-7

IUPAC Name: sodium nonyl sulfate
Synonyms: sodium nonyl sulfate
Molecular Weight (g/mol): 246.3
Molecular Formula: C9H19NaO4S
InChi Key: FTWCSAMTIKSPAT-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCOS([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results