man

Test

CAS RN 1072952-53-4

BOClHOOH

CAS RN 1072952-53-4

IUPAC Name: (3-chloro-4-formylphenyl)boronic acid
Synonyms: 3-chloro-4-formylphenylboronic acid
Molecular Weight (g/mol): 184.38
Molecular Formula: C7H6BClO3
InChi Key: KDIYVLXLPNHJRZ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Cl)=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results