man

Test

CAS RN 1073-72-9

H3CSOH

CAS RN 1073-72-9

IUPAC Name: 4-(methylsulfanyl)phenol
Synonyms: 4-(methylthio)-phenol
Molecular Weight (g/mol): 140.2
Molecular Formula: C7H8OS
InChi Key: QASBCTGZKABPKX-UHFFFAOYSA-N
SMILES: CSC1=CC=C(O)C=C1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)