man

Test

CAS RN 1073354-84-3

BOOH3CCH3H3CH3CNHON

CAS RN 1073354-84-3

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-dihydropyrimidin-2-one
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrimidin-2-one
Molecular Weight (g/mol): 222.05
Molecular Formula: C10H15BN2O3
InChi Key: JRCSRFGCQSLWNZ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CNC(=O)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results