man

Test

CAS RN 1074-36-8

HOOSH

CAS RN 1074-36-8

IUPAC Name: 4-sulfanylbenzoic acid
Synonyms: 2-mercaptobenzoic acid
Molecular Weight (g/mol): 154.18
Molecular Formula: C7H6O2S
InChi Key: LMJXSOYPAOSIPZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(S)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results