man

Test

CAS RN 1074-92-6

CH3H3CCH3H3C

CAS RN 1074-92-6

IUPAC Name: 1-tert-butyl-2-methylbenzene
Synonyms: 1-tert-butyl-2-methylbenzene
Molecular Weight (g/mol): 148.25
Molecular Formula: C11H16
InChi Key: AXHVNJGQOJFMHT-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results