CAS RN 107558-48-5
CAS RN 107558-48-5
IUPAC Name:
4-[(pyridin-4-yl)methyl]-1,2-dihydrophthalazin-1-one
Synonyms:
4-(pyridin-4-ylmethyl)-2H-phthalazin-1-one
Molecular Weight (g/mol):
237.26
Molecular Formula:
C14H11N3O
InChi Key:
NIQMWTLRDNQDIA-UHFFFAOYSA-N
SMILES:
O=C1NN=C(CC2=CC=NC=C2)C2=CC=CC=C12
1
–
1
of
1
results
1
4-(4-Pyridylmethyl)-1(2H)-phthalazinone, 96%, Thermo Scientific™
CAS: 107558-48-5 Molecular Formula: C14H11N3O Molecular Weight (g/mol): 237.262 MDL Number: MFCD00443664 InChI Key: NIQMWTLRDNQDIA-UHFFFAOYSA-N Synonym: 4-4-pyridylmethyl-1 2h-phthalazinone,4-pyridin-4-ylmethyl phthalazin-1 2h-one,4-pyridin-4-ylmethyl-2h-phthalazin-1-one,4-4-pyridinylmethyl-1 2h-phthalazinone,1 2h-phthalazinone,4-4-pyridinylmethyl,4-pyridin-4-ylmethyl-1,2-dihydrophthalazin-1-one,enamine_000224,cambridge id 5237055,4-4-pyridylmethyl-phthalazinone,4-pyridin-4-ylmethyl phthalazin-1-ol PubChem CID: 789954 IUPAC Name: 4-(pyridin-4-ylmethyl)-2H-phthalazin-1-one SMILES: C1=CC=C2C(=C1)C(=NNC2=O)CC3=CC=NC=C3