CAS RN 107818-20-2
CAS RN 107818-20-2
IUPAC Name:
4H,7H-thieno[3,2-b]pyridin-7-one
Synonyms:
4H-thieno[3,2-b]pyridin-7-one
Molecular Weight (g/mol):
151.18
Molecular Formula:
C7H5NOS
InChi Key:
AACVULYSNJAKEQ-UHFFFAOYSA-N
SMILES:
O=C1C=CNC2=C1SC=C2
1
–
1
of
1
results
1
Thieno[3,2-b]pyridin-7-ol, 96%, Thermo Scientific Chemicals
CAS: 107818-20-2 Molecular Formula: C7H5NOS Molecular Weight (g/mol): 151.183 MDL Number: MFCD01863636 InChI Key: AACVULYSNJAKEQ-UHFFFAOYSA-N Synonym: thieno 3,2-b pyridin-7-ol,thieno 3,2-b pyridin-7 4h-one,7-hydroxythieno 3,2-b pyridine,4h-thieno 3,2-b pyridin-7-one,thiopheno 3,2-b pyridin-7-ol,thieno 3 2-b pyridin-7-ol,pubchem22356,acmc-2098xl,ksc173k8p PubChem CID: 5074765 IUPAC Name: 4H-thieno[3,2-b]pyridin-7-one SMILES: C1=CNC2=C(C1=O)SC=C2