man

Test

CAS RN 108-12-3

CH3H3CClO

CAS RN 108-12-3

IUPAC Name: 3-methylbutanoyl chloride
Synonyms: butanoyl chloride, 3-methyl-
Molecular Weight (g/mol): 120.58
Molecular Formula: C5H9ClO
InChi Key: ISULZYQDGYXDFW-UHFFFAOYSA-N
SMILES: CC(C)CC(Cl)=O
Boiling Point 
  • (6)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)