man

Test

CAS RN 108-47-4

CH3NH3C

CAS RN 108-47-4

IUPAC Name: 2,4-dimethylpyridine
Synonyms: 2,4-dimethylpyridine
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: JYYNAJVZFGKDEQ-UHFFFAOYSA-N
SMILES: CC1=CC=NC(C)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)