man

Test

CAS RN 108-70-3

ClClCl

CAS RN 108-70-3

IUPAC Name: 1,3,5-trichlorobenzene
Synonyms: 1,3,5-trichloro-benzene s-trichlorobenzene sym-trichlorobenzene
Molecular Weight (g/mol): 181.44
Molecular Formula: C6H3Cl3
InChi Key: XKEFYDZQGKAQCN-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=CC(Cl)=C1
Quantity 
  • (3)
  • (3)
Type 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)